Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

614904

Sigma-Aldrich

Methane-d3-thiol

98 atom % D

Synonym(s):

Methyl-d3 mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3SH
CAS Number:
Molecular Weight:
51.13
MDL number:
UNSPSC Code:
12352105
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

mass shift

M+3

SMILES string

[2H]C([2H])([2H])S

InChI

1S/CH4S/c1-2/h2H,1H3/i1D3

InChI key

LSDPWZHWYPCBBB-FIBGUPNXSA-N

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Standard 5 g quantity is packaged in a 25 mL Sure/Pac cylinder with stainless steel 1/4" male NPT valve. Nominal gas pressure at 21ºC is 15 psig for 5 g. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel hose adapter Z14,683-8 or stainless steel body mini gas regulator Z51,354-7 is recommended.

Legal Information

Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jared A Wiig et al.
Nature communications, 6, 8034-8034 (2015-08-12)
Carbide insertion plays a pivotal role in the biosynthesis of M-cluster, the cofactor of nitrogenase. Previously, we proposed a carbide insertion pathway involving methyltransfer from SAM to a FeS precursor and hydrogen abstraction from this methyl group that initiates the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service