Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

609390

Sigma-Aldrich

Ammonium-15N hydroxide solution

~14 N in H2O, 98 atom % 15N

Synonym(s):

15N Labeled ammonium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
15NH4OH/H2O soln.
CAS Number:
Molecular Weight:
36.04
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

concentration

~14 N in H2O

mass shift

M+1

storage temp.

2-8°C

SMILES string

[15NH3].O

InChI

1S/H3N.H2O/h1H3;1H2/i1+1;

InChI key

VHUUQVKOLVNVRT-YTBWXGASSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service