Skip to Content
MilliporeSigma

609331

Ammonia-15N solution

98 atom % 15N, 2 M in methanol

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
15NH3
Molecular Weight:
18.02
UNSPSC Code:
12352116
PubChem Substance ID:
MDL number:
Isotopic purity:
98 atom % 15N
Mass shift:
M+1


isotopic purity

98 atom % 15N

concentration

2 M in methanol

mass shift

M+1

SMILES string

[15NH3]

InChI

1S/H3N/h1H3/i1+1

InChI key

QGZKDVFQNNGYKY-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.


Still not finding the right product?

Explore all of our products under Ammonia-15N solution


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library