Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

607118

Sigma-Aldrich

Methylamine-13C,15N

99 atom % 13C, 98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH315NH2
CAS Number:
Molecular Weight:
33.04
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % 15N

technique(s)

protein expression: suitable

bp

-6.3 °C (lit.)

mp

-93 °C (lit.)

mass shift

M+2

SMILES string

[13CH3][15NH2]

InChI

1S/CH5N/c1-2/h2H2,1H3/i1+1,2+1

InChI key

BAVYZALUXZFZLV-ZDOIIHCHSA-N

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Flam. Gas 1A - Press. Gas Compr. Gas - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

<-22.0 °F

Flash Point(C)

< -30 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service