Sign In to View Organizational & Contract Pricing.
About This Item
Linear Formula:
CD3CD2NH2
CAS Number:
Molecular Weight:
50.11
UNSPSC Code:
12352202
PubChem Substance ID:
MDL number:
Isotopic purity:
99 atom % D
Mass shift:
M+5
isotopic purity
99 atom % D
Quality Segment
mass shift
M+5
SMILES string
[2H]C([2H])([2H])C([2H])([2H])N
InChI
1S/C2H7N/c1-2-3/h2-3H2,1H3/i1D3,2D2
InChI key
QUSNBJAOOMFDIB-ZBJDZAJPSA-N
Packaging
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Gas 1 - Press. Gas Liquefied gas - STOT SE 3
target_organs
Respiratory system
Storage Class
2A - Gases
wgk
WGK 1
flash_point_f
-34.6 °F - closed cup
flash_point_c
-37.00 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


