Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

605751

Sigma-Aldrich

Potassium palmitate-13C16

99 atom % 13C, 99% (CP)

Synonym(s):

Hexadecanoic acid-13C16 potassium salt, Palmitic acid-13C16 potassium salt, Potassium hexadecanoate-13C16

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3(13CH2)1413CO2K
CAS Number:
Molecular Weight:
310.40
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

solid

mp

285 °C (lit.)

mass shift

M+16

SMILES string

[K+].[13CH3][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13C]([O-])=O

InChI

1S/C16H32O2.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18;/h2-15H2,1H3,(H,17,18);/q;+1/p-1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1;

InChI key

MQOCIYICOGDBSG-SJIUKAAASA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Rachel J Perry et al.
Cell, 160(4), 745-758 (2015-02-11)
Impaired insulin-mediated suppression of hepatic glucose production (HGP) plays a major role in the pathogenesis of type 2 diabetes (T2D), yet the molecular mechanism by which this occurs remains unknown. Using a novel in vivo metabolomics approach, we show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service