Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

603147

Sigma-Aldrich

Water-18O

80 atom % 18O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H218O
CAS Number:
Molecular Weight:
20.02
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:

description

Normalized with respect to hydrogen

isotopic purity

80 atom % 18O

mol wt

19.62 g/mol

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.11 g/mL at 20 °C (lit.)

mass shift

M+2

SMILES string

[18OH2]

InChI

1S/H2O/h1H2/i1+2

InChI key

XLYOFNOQVPJJNP-NJFSPNSNSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service