Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

593427

Sigma-Aldrich

2-Hydroxy-5-methylpyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

183-187 °C (lit.)

SMILES string

Cc1ccc(O)nc1

InChI

1S/C6H7NO/c1-5-2-3-6(8)7-4-5/h2-4H,1H3,(H,7,8)

InChI key

SOHMZGMHXUQHGE-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Emma Kirkman-Davis et al.
Dalton transactions (Cambridge, England : 2003), 49(39), 13693-13703 (2020-10-01)
A family of pyrazine-bridged, linear chain complexes of Cu(ii) of the formula [CuL2(H2O)2(pz)](ClO4)2 [pz = pyrazine; L = n-methyl-2(1H)-pyridone, n = 3 (1), 5 (2), and 6 (3)] has been prepared. Single-crystal X-ray diffraction shows six-coordinate, pyrazine-bridged chains with trans-pairs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service