Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

588032

Sigma-Aldrich

Dibenzothiophene-d8

98 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D8S
CAS Number:
Molecular Weight:
192.31
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

98% (CP)

form

solid

bp

323-333 °C

mp

97-100 °C

mass shift

M+8

SMILES string

[2H]c1c([2H])c([2H])c2c(sc3c([2H])c([2H])c([2H])c([2H])c23)c1[2H]

InChI

1S/C12H8S/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8H/i1D,2D,3D,4D,5D,6D,7D,8D

InChI key

IYYZUPMFVPLQIF-PGRXLJNUSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Badreddine Barhoumi et al.
Marine environmental research, 101, 184-195 (2014-08-12)
In this study, biological responses and contaminant levels in biological tissues were investigated in grass goby fish specimens (Zosterisessor ophiocephalus) collected from five stations in a moderately polluted ecosystem, namely the Bizerte lagoon on the north coast of Tunisia. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service