Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

579025

Sigma-Aldrich

Acetanilide-d8

98 atom % D

Synonym(s):

N-(Phenyl-2,3,4,5,6-d5)acetamide-2,2,2-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CONHC6D5
CAS Number:
Molecular Weight:
143.21
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

form

solid

bp

304 °C (lit.)

mp

113-115 °C (lit.)

density

1.290 g/mL at 25 °C

mass shift

M+8

SMILES string

[2H]c1c([2H])c([2H])c(NC(=O)C([2H])([2H])[2H])c([2H])c1[2H]

InChI

1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)/i1D3,2D,3D,4D,5D,6D

InChI key

FZERHIULMFGESH-JGUCLWPXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service