Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

749079

Sigma-Aldrich

Oleic acid-1,2,3,7,8-13C5

99 atom % 13C, 99% (CP)

Synonym(s):

cis-9-Octadecenoic acid-1,2,3,7,8-13C5, Elainic acid-1,2,3,7,8-13C5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(13CH2)2(CH2)3(13CH2)213COOH
CAS Number:
Molecular Weight:
287.42
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

liquid

bp

194-195 °C/1.2 mmHg (lit.)

mp

13-14 °C (lit.)

density

0.910 g/mL at 25 °C (lit.)

mass shift

M+5

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/[13CH2][13CH2]CCC[13CH2][13CH2][13C](O)=O

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9-/i11+1,12+1,16+1,17+1,18+1

InChI key

ZQPPMHVWECSIRJ-TWKVKCSCSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service