Skip to Content
MilliporeSigma
All Photos(1)

Documents

56760

Sigma-Aldrich

Triethoxy-3-(2-imidazolin-1-yl)propylsilane

≥97.0% (NT)

Synonym(s):

1-(3-Triethoxysilylpropyl)-2-imidazoline, 3-(2-Imidazolin-1-yl)propyltriethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H26N2O3Si
CAS Number:
Molecular Weight:
274.43
Beilstein:
8987333
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (NT)

refractive index

n20/D 1.452

density

1.005 g/mL at 20 °C (lit.)

SMILES string

CCO[Si](CCCN1CCN=C1)(OCC)OCC

InChI

1S/C12H26N2O3Si/c1-4-15-18(16-5-2,17-6-3)11-7-9-14-10-8-13-12-14/h12H,4-11H2,1-3H3

InChI key

WBUSESIMOZDSHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Additive for inorganic modification of block copolymers

Catalyst for condensation reactions

Modifier for organically modified layered silicates

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Zhang et al.
Macromolecular rapid communications, 40(11), e1900067-e1900067 (2019-04-04)
A particle mold synthesis of 2D Janus nanomaterials is proposed by crosslinking of copolymer self-assembled monolayers confined within the mold domains. Onto the silica (SiO2 ) particle surface, mold domains with functional groups such as imidazole are generated. The model
Mehdi Abedi et al.
International journal of nanomedicine, 15, 2617-2631 (2020-05-06)
As widely used chemotherapeutic agents, platinum compounds have several therapeutic challenges, such as drug resistance and adverse effects. Theranostic systems, macromolecular or colloidal therapeutics with companion diagnostics, not only address controlled drug delivery but also enable real-time monitoring of tumor sites.
Youngdo Kim et al.
Scientific reports, 9(1), 10293-10293 (2019-07-18)
In this study, we synthesized molecularly imprinted mesoporous organosilica (MIMO) in the presence of a new precursor having a zwitterionic functional group and an imprint molecule, namely, glyphosate (MIMO-z). The precursor-glyphosate complex engaged in a typical base-catalyzed sol-gel reaction and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service