Skip to Content
MilliporeSigma
Triethoxy-3-(2-imidazolin-1-yl)propylsilane ≥97.0% (NT)

Triethoxy-3-(2-imidazolin-1-yl)propylsilane

Triethoxy-3-(2-imidazolin-1-yl)propylsilane ≥97.0% (NT)
Synonym(s):
1-(3-Triethoxysilylpropyl)-2-imidazoline, 3-(2-Imidazolin-1-yl)propyltriethoxysilane
Empirical Formula (Hill Notation):
C12H26N2O3Si
CAS Number:
Molecular Weight:
274.43
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing