Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

553743

Sigma-Aldrich

4,4-Pentamethylene-2-pyrrolidinone

96%

Synonym(s):

3-Azaspiro[4.5]decan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO
CAS Number:
Molecular Weight:
153.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
96%

Quality Level

Assay

96%

form

solid

mp

84-89 °C (lit.)

SMILES string

O=C1CC2(CCCCC2)CN1

InChI

1S/C9H15NO/c11-8-6-9(7-10-8)4-2-1-3-5-9/h1-7H2,(H,10,11)

InChI key

JAWPQJDOQPSNIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4-Pentamethylene-2-pyrrolidinone (2-azaspiro[4.5]decan-3-one, gabapentin-lactam) is a degradation product of gabapentin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annalisa Cutrignelli et al.
International journal of pharmaceutics, 332(1-2), 98-106 (2006-10-31)
The aim of this study was to gain insights into the role played by some excipients on the stability of gabapentin 1 and baclofen 2 which can undergo degradation giving rise to the corresponding lactams 2-azaspiro[4.5]decan-3-one 3 and 4-(4-chlorophenyl)-2-pyrrolidone 4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service