Skip to Content
MilliporeSigma
All Photos(1)

Documents

552542

Sigma-Aldrich

(R)-(+)-2-(Diphenylmethyl)pyrrolidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N
CAS Number:
Molecular Weight:
237.34
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

optical activity

[α]20/D +3.0°, c = 1% in chloroform

refractive index

n20/D 1.5870 (lit.)

density

1.062 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CN[C@H](C1)C(c2ccccc2)c3ccccc3

InChI

1S/C17H19N/c1-3-8-14(9-4-1)17(16-12-7-13-18-16)15-10-5-2-6-11-15/h1-6,8-11,16-18H,7,12-13H2/t16-/m1/s1

InChI key

OXOBKZZXZVFOBB-MRXNPFEDSA-N

Application

Used as excellent chiral solvating agents to determine the enantiomeric composition of chiral carboxylic acids directly by NMR analysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bailey, D. J. et al.
Tetrahedron Asymmetry, 8, 149-149 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service