Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

677183

Sigma-Aldrich

(S)-(–)-α,α-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether

95%

Synonym(s):

(2S)-2-[Diphenyl[(trimethylsilyl)oxy]methyl]pyrrolidine, (S)-α,α-Diphenylprolinol trimethylsilyl ether, (S)-2-[Diphenyl(trimethylsilanyloxy)methyl]pyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27NOSi
CAS Number:
Molecular Weight:
325.52
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

optical activity

[α]22/D -50°, c = 1 in chloroform

refractive index

n20/D 1.5513

density

1.0459 g/mL at 25 °C

functional group

phenyl

storage temp.

2-8°C

SMILES string

C[Si](C)(C)OC([C@@H]1CCCN1)(c2ccccc2)c3ccccc3

InChI

1S/C20H27NOSi/c1-23(2,3)22-20(19-15-10-16-21-19,17-11-6-4-7-12-17)18-13-8-5-9-14-18/h4-9,11-14,19,21H,10,15-16H2,1-3H3/t19-/m0/s1

InChI key

RSUHWMSTWSSNOW-IBGZPJMESA-N

Application

Organocatalyst for 3-component reactions, and Robinson annulation of α,β-unsaturated aldehydes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bor-Cherng Hong et al.
The Journal of organic chemistry, 72(22), 8459-8471 (2007-10-09)
The highly enantioselective organocatalytic Robinson annulation of alpha,beta-unsaturated aldehydes was achieved, catalyzed by l-proline and trialkylamines and providing the formal [4 + 2] cycloaddition adducts. Additionally, in some examples in the catalysis with diarylpyrrolinol silyl ethers, the reactions afforded the
Dieter Enders et al.
Nature, 441(7095), 861-863 (2006-06-17)
Efficient and elegant syntheses of complex organic molecules with multiple stereogenic centres continue to be important in both academic and industrial laboratories. In particular, catalytic asymmetric multi-component 'domino' reactions, used during total syntheses of natural products and synthetic building blocks
Chemical and Engineering News, June (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service