Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Linear Formula:
FC6H2(OH)(NO2)COCH3
CAS Number:
Molecular Weight:
199.14
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Assay:
98%
Pricing and availability is not currently available.
Quality Segment
assay
98%
mp
87-90 °C (lit.)
functional group
fluoro, ketone, nitro
SMILES string
CC(=O)c1cc(F)cc(c1O)[N+]([O-])=O
InChI
1S/C8H6FNO4/c1-4(11)6-2-5(9)3-7(8(6)12)10(13)14/h2-3,12H,1H3
InChI key
YVTPUCHIOSGVSH-UHFFFAOYSA-N
General description
5′-Fluoro-2′-hydroxy-3′-nitroacetophenone, also known as 1-(5-fluoro-2-hydroxy-3-nitrophenyl)ethanone, is an aromatic hydroxy ketone. It can be synthesized from 5-fluoro-2-hydroxy-acetophenone via nitration.
Application
5′-Fluoro-2′-hydroxy-3′-nitroacetophenone may be used to prepare 1-(3-amino-5-fluoro-2-hydroxyphenyl)ethanone via catalytic hydrogenation.
1 of 1
This Item | |||
|---|---|---|---|
| description 98% | description 99% | description 98 atom % D | description PESTANAL®, analytical standard |
| assay 98% | assay 99% | assay - | assay ≥98.0% (HPLC) |
| Quality Level 100 | Quality Level 100 | Quality Level 200 | Quality Level 100 |
| mp 87-90 °C (lit.) | mp 99-101 °C (lit.) | mp 14-18 °C (lit.) | mp - |
| functional group ketone, nitro, fluoro | functional group nitro, ketone, chloro | functional group - | functional group - |
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




