Skip to Content
MilliporeSigma
All Photos(1)

Documents

193968

Sigma-Aldrich

4′-Chloro-3′-nitroacetophenone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)COCH3
CAS Number:
Molecular Weight:
199.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

99-101 °C (lit.)

SMILES string

CC(=O)c1ccc(Cl)c(c1)[N+]([O-])=O

InChI

1S/C8H6ClNO3/c1-5(11)6-2-3-7(9)8(4-6)10(12)13/h2-4H,1H3

InChI key

YEVPHFIFGUWSMG-UHFFFAOYSA-N

General description

4′-Chloro-3′-nitroacetophenone is the intermediate formed during the synthesis of 4-chloro-3-nitrostyrene. It participates in deamination reaction of 4-chloro-5- and -3-nitro-2-aminoacetophanones.

Application

4′-Chloro-3′-nitroacetophenone was used to prepare starting reagent for the synthesis of 6- and 7-acetyl-3-methyl-2-quinoxalinecarboxamide1,4-dioxides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 4-chloro-3-nitrostyrene.
Strantzalis N, et al.
Polym. Bull., 15(5), 431-438 (1986)
Cinnolines; the preparation of 4-chloro-2-amino-acetophenone and related 4-hydroxycinnolines.
C M ATKINSON et al.
Journal of the Chemical Society, 232-237 (1947-02-01)
J P Dirlam et al.
Journal of medicinal chemistry, 21(5), 483-485 (1978-05-01)
The synthesis, separation, and structure determination of 6- and 7-acetyl--3-methyl-2-quinoxalinecarboxamide 1,4-dioxides are reported together with a comparison of their antibacterial activity. The structural assignment of these 6- and7-acetyl isomers was based on NMR analysis of related mono-N-oxide derivatives, which were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service