Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

543349

Sigma-Aldrich

IR-806

Dye content 90 %

Synonym(s):

2-[2-[2-chloro-3-[2-[1,3-dihydro-3,3-dimethyl-1-(4-sulfobutyl)-2H-indol-2-ylidene]-ethylidene]-1-cyclopenten-1-yl]-ethenyl]-3,3-dimethyl-1-(4-sulfobutyl)-3H-indolium hydroxide, inner salt sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H44ClN2NaO6S2
Molecular Weight:
735.33
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Dye content, 90%

mp

214-218 °C (lit.)

λmax

806 nm

SMILES string

[Na+].CC1(C)\C(=C\C=C2/CCC(\C=C\C3=[N+](CCCCS([O-])(=O)=O)c4ccccc4C3(C)C)=C2Cl)N(CCCCS([O-])(=O)=O)c5ccccc15

InChI

1S/C37H45ClN2O6S2.Na/c1-36(2)29-13-5-7-15-31(29)39(23-9-11-25-47(41,42)43)33(36)21-19-27-17-18-28(35(27)38)20-22-34-37(3,4)30-14-6-8-16-32(30)40(34)24-10-12-26-48(44,45)46;/h5-8,13-16,19-22H,9-12,17-18,23-26H2,1-4H3,(H-,41,42,43,44,45,46);/q;+1/p-1

InChI key

MIOUAELAUIYKFC-UHFFFAOYSA-M

General description

IR-806 is a near infrared (NIR) cyanine dye with an absorption peak in the range of 730-800 nm. It is a water soluble dye that is commonly used as an active medium in NIR dye lasers. It is a photo stable dye that is useful in biomedical applications.

Application

IR-806 can be used for a variety of applications such as dye sensitized solar cells (DSSCs), bio-imaging, and optical displays.

Features and Benefits

Water soluble, laser and near-IR dye

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new water-soluble near-infrared croconium dye
Song X and Foley JW
Dyes and Pigments, 78(1), 60-64 (2008)
Dye-sensitized core/active shell upconversion nanoparticles for optogenetics and bioimaging applications
Wu X, et al.
ACS Nano, 10(1), 1060-1066 (2016)
Large assembly formation via a two-step process in a chromonic liquid crystal
Mills EA, et al.
The Journal of Physical Chemistry B, 116(45), 13506-13515 (2012)
Broadband dye-sensitized upconversion of near-infrared light
Zou W, et al.
Nature Photonics, 6(8), 560-560 (2012)
Multiple hybridized resonances of IR-806 chromonic molecules strongly coupled to Au nanorods
Zhang Y, et al.
Nanoscale, 7(18), 8503-8509 (2015)

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service