Skip to Content
MilliporeSigma

90487

NIR 5e

≥80% (HPCE), suitable for fluorescence, BioReagent

Synonym(s):

1,1′-Bis(3-isothiocyanatopropyl)-11-chloro-4,5:4′,5′-dibenzo-3,3,3′,3′-tetramethyl-10,12-trimethylenindotricarbocyanine bromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C46H46BrClN4S2
CAS Number:
Molecular Weight:
834.37
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352108
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

assay

≥80% (HPCE)

form

crystals

fluorescence

λex 822 nm; λem 844 nm in methanol

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[Br-].CC1(C)\C(=C/C=C2\CCCC(\C=C\C3=[N+](CCCN=C=S)c4ccc5ccccc5c4C3(C)C)=C2Cl)N(CCCN=C=S)c6ccc7ccccc7c16

InChI

1S/C46H46ClN4S2.BrH/c1-45(2)40(50(28-10-26-48-30-52)38-22-18-32-12-5-7-16-36(32)42(38)45)24-20-34-14-9-15-35(44(34)47)21-25-41-46(3,4)43-37-17-8-6-13-33(37)19-23-39(43)51(41)29-11-27-49-31-53;/h5-8,12-13,16-25H,9-11,14-15,26-29H2,1-4H3;1H/q+1;/p-1

InChI key

NTQGWDKAUSUQIS-UHFFFAOYSA-M

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
90487-1MG-F04061823359672
90487-5MG-F04061832533681