Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

538264

Sigma-Aldrich

Butyronitrile

≥99%

Synonym(s):

Propyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CN
CAS Number:
Molecular Weight:
69.11
Beilstein:
1361452
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.4 (vs air)

vapor pressure

23 mmHg ( 25 °C)

Assay

≥99%

autoignition temp.

910 °F

refractive index

n20/D 1.384 (lit.)

bp

115-117 °C (lit.)

mp

−112 °C (lit.)

solubility

DMF: miscible(lit.)
alcohol: miscible(lit.)
diethyl ether: miscible(lit.)
water: very slightly soluble(lit.)

density

0.794 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

CCCC#N

InChI

1S/C4H7N/c1-2-3-4-5/h2-3H2,1H3

InChI key

KVNRLNFWIYMESJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Butyronitrile (Propyl cyanide, Butane nitrile, BN) is a linear cyanide. Butyronitrile (BN) glasses were fabricated by vapor deposition. The relationship between molar volume and deposition temperature for BN glasses has been investigated. Its reaction with high-temperature water (Temperature = 330°C and Pressure range = 128-2600bar) has been reported to afford butanamide, butyric acid and ammonia. It undergoes selective reduction in the presence of LiOH promoted Co-based Raney-catalysts to afford n-butylamine.

Application

Butyronitrile is suitable for use in a study to investigate the effect of stereochemical factors on intervalence charge transfer (IT). It may be used in electrolyte composition in dye-sensitized solar cells.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tailoring Raney-catalysts for the selective hydrogenation of butyronitrile to n-butylamine.
Chojecki A, et al.
J. Catal., 245(1), 237-248 (2007)
Stereochemical influences on intervalence charge transfer in homodinuclear complexes of ruthenium.
D M D'Alessandro et al.
Inorganic chemistry, 40(26), 6841-6844 (2001-12-12)
Effect of pressure on the rate of butyronitrile hydrolysis in high-temperature water.
Iyer SD and Klein MT.
Journal of Supercritical Fluids, 10(3), 191-200 (1997)
Frédéric Sauvage et al.
Journal of the American Chemical Society, 133(33), 13103-13109 (2011-06-28)
We elaborated a new electrolyte composition, based on butyronitrile solvent, that exhibits low volatility for use in dye-sensitized solar cells. The strong point of this new class of electrolyte is that it combines high efficiency and excellent stability properties, while
Hideyuki Nakayama et al.
The journal of physical chemistry. B, 117(35), 10311-10319 (2013-08-08)
Molar volumes (Vm) of vapor-deposited ethylcyclohexane (ECH) and butyronitrile (BN, sometimes called butanenitrile) glasses were studied as a function of deposition temperature (Td). ECH glasses deposited at Td sufficiently below their glass-transition temperature (Tg) exhibited changes in Vm on heating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service