Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

532770

Sigma-Aldrich

1H,1H,2H,2H-Perfluorohexan-1-ol

97%

Synonym(s):

3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)3(CH2)2OH
CAS Number:
Molecular Weight:
264.09
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.314 (lit.)

bp

140-143 °C (lit.)

density

1.59 g/mL at 25 °C (lit.)

application(s)

PFAS testing

SMILES string

OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C6H5F9O/c7-3(8,1-2-16)4(9,10)5(11,12)6(13,14)15/h16H,1-2H2

InChI key

JCMNMOBHVPONLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.9 °F - closed cup

Flash Point(C)

73.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J F Ayala-Cabrera et al.
Journal of chromatography. A, 1609, 460463-460463 (2019-08-27)
In this work, the suitability of gas chromatography-mass spectrometry (GC-MS) and liquid chromatography-tandem mass spectrometry (LC-MS/MS) for the multi-class determination of different families of neutral per- and polyfluoroalkyl substances (PFASs), such as fluorotelomer olefins (FTOs), alcohols (FTOHs) and fluorooctanesulfonamides (FOSAs)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service