446831
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluoro-1-decanol
98%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
98%
form
solid
bp
192 °C/735 mmHg (lit.)
mp
87-90 °C (lit.)
functional group
fluoro
hydroxyl
SMILES string
OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
InChI
1S/C10H3F19O/c11-2(12,1-30)3(13,14)4(15,16)5(17,18)6(19,20)7(21,22)8(23,24)9(25,26)10(27,28)29/h30H,1H2
InChI key
NIRPXSQCRWXHNZ-UHFFFAOYSA-N
General description
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluoro-1-decanol is a fluorotelomer alcohol (FTOH).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Biological & pharmaceutical bulletin, 30(7), 1358-1359 (2007-07-03)
The present study demonstrates the estrogenic effects of fluorotelomer alcohols (FTOHs). In a yeast two-hybrid assay, treatment with 1H,1H,2H,2H-perfluorooctan-1-ol (6:2 FTOH), 1H,1H,2H,2H-perfluoro-decan-1-ol (8:2 FTOH) and 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluoro-1-decanol (NFDH) showed a dose-dependent interaction between the human estrogen receptor (hER) isoforms hERalpha or
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service