Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

532304

Sigma-Aldrich

2-Chlorotrityl chloride, polymer-bound

100-200 mesh, extent of labeling: 1.0-1.5 mmol/g

Synonym(s):

2,α-Dichlorobenzhydryl-polystyrene crosslinked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
1983914
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

1.0-1.5 mmol/g

particle size

100-200 mesh

application(s)

peptide synthesis

SMILES string

Clc1ccccc1C(Cl)(c2ccccc2)c3ccccc3

InChI

1S/C19H14Cl2/c20-18-14-8-7-13-17(18)19(21,15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14H

InChI key

JFLSOKIMYBSASW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Acid labile resin used in Fmoc-based solid phase peptide synthesis.
  • Mild acidic cleavage conditions lead to the release of the peptide acid where fully protected peptides can be released if desired.
  • 2-Chlorotrityl chloride resins prevent racemization of the first amino acid and are thus very useful when racemic mixtures are forming (common with residues such as His or Cys).
  • This resin also prevents diketopiperazide formation, which can be an issue with proline C-terminal peptide sequences.

Use:
  • Attachment of the first amino acid residue is effected by stirring the resin, the protected amino acid, and excess diisopropylethylamine (DIEA) in dichloromethane.
  • Cleavage of the final protected peptide fragment is achieved under very mild conditions using either acetic acid/trifluoroethanol (TFE)/dichloromethane (1:1:8; v/v/v), hexafluoroisopropanol (HFIP)/dichloromethane (1:4; v/v) or simply 0.5% trifluoroacetic acid/dichloromethane (v/v).
  • Higher concentrations of TFA can be used if retention of peptide side chaing protecting groups is unimportant. Note that trityl chloride is moisture-sensitive, and, therefore, should be stored and handled appropriately.
  • If the resin becomes deactivated, treatment with acetyl chloride or SOCl2 in toluene before use is recommended to restore its activity.

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service