Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.55012

Sigma-Aldrich

Trityl chloride resin

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352101

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-30°C

General description

An acid-labile resin for the solid phase immobilization of alcohols [1,2,3,4,5], amines [4, 6,7,8,9,10,11,12], sugars [13], O- and N-linked hydroxylamines [14,15], imidazoles [16] and carboxylic acids [16]. Release of these functionalities is generally achieved using 1-5% TFA in DCM containing 5% TIS. Amines can also be cleaved with 30% HFIP in DCM [4]. This resin, although very acid sensitive, can also be used in peptide synthesis [17].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] C. C. Leznoff (1978) Acc. Chem. Res., 11, 327.
[2] C. Chen, et al. (1994) J. Am. Chem. Soc., 116, 2661.
[3] Z. Li & A. Ganesan (1998) Syn. Lett., 405.
[4] P. Athanassopoulos, et al., Poster 316 presented at the 24thEuropean Peptide Symposium, Edinburgh, 1996.
[5] K. Barlos, et al. (1988) Liebigs Ann. Chem., 1079.
[6] K. Barlos, et al. in ′Peptides 1992, Proc. 22nd European Peptide Symposium′, C. H. Schneider & A. N. Eberle (Eds), ESCOM, Leiden,1993, pp. 281.
[7] M. Schuster, et al. (1996) Angew. Chem. Int. Ed. Engl., 35, 1979.
[8] P. Garibay, et al. (1998) Tetrahedron Lett., 39, 2207.
[9] Y. Guan, et al. (2000) J. Comb. Chem., 2, 297.
[10] S. Manku, et al. (2001) J. Org. Chem., 66, 874.
[11] D. Jönsson, et al. (2001) Tetrahedron Lett., 42, 6953.
[12] C. A. Olsen, et al. (2003) Org. Lett., 5, 4183.
[13] F. Peri, et al. (2000) Tetrahedron Lett., 41, 8587.
[14] U. Bauer, et al. (1997) Tetrahedron Lett., 38, 7233.
[15] O. Kinzel, et al. (2003) J. Peptide Sci., 9, 375.
[16] D. P. Matthews, et al. (2000) J Comb. Chem., 2,19.
[17] K. Barlos, et al. (1989) Tetrahedron Lett., 30, 3943.

Linkage

Replaces: 01-64-0074

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Ala-Leu loaded resin): 1.1 - 1.7 mmol/g
Swelling Volume (in CH₂Cl₂): lot specific result
The polymer matrix is copoly (styrene-1 % DVB), 200 - 400 mesh

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service