Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

530549

Sigma-Aldrich

Ethylene glycol-d6

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DOCD2CD2OD
CAS Number:
Molecular Weight:
68.10
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.1 (vs air)

vapor pressure

0.08 mmHg

isotopic purity

98 atom % D

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.431 (lit.)

bp

196-198 °C (lit.)

mp

-13 °C (lit.)

density

1.220 g/mL at 25 °C

mass shift

M+6

SMILES string

[2H]OC([2H])([2H])C([2H])([2H])O[2H]

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2/i1D2,2D2,3D,4D

InChI key

LYCAIKOWRPUZTN-UFSLNRCZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylene glycol-d6 is a deuterated NMR solvent useful in NMR-based research and analyses. Its infrared spectrum has been recorded.

Application

Ethylene glycol-d6 may be used as starting material for the synthesis of oxiran-d4.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The mechanism of the cationic cyclooligomerization of ethylene-oxide and the concomitant degradation of the crown ether products formed.
Dale J and Daasvatn K.
Acta Chemica Scandinavica. Series B, 34(5), 327-342 (1980)
Infrared spectra and molecular vibrations of ethylene glycol and deuterated derivatives.
Matsuura H and Miyazawa T.
Bulletin of the Chemical Society of Japan, 40(1), 85-94 (1967)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service