Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

277592

Sigma-Aldrich

9-Vinylcarbazole

98%

Synonym(s):

9-Vinyl-9H-carbazole, N-Ethenylcarbazole, N-Vinylcarbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11N
CAS Number:
Molecular Weight:
193.24
Beilstein:
132988
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

bp

154-155 °C/3 mmHg (lit.)

mp

60-65 °C (lit.)

SMILES string

C=Cn1c2ccccc2c3ccccc13

InChI

1S/C14H11N/c1-2-15-13-9-5-3-7-11(13)12-8-4-6-10-14(12)15/h2-10H,1H2

InChI key

KKFHAJHLJHVUDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Merve Kara et al.
Macromolecular rapid communications, 36(23), 2070-2075 (2015-09-12)
A new phenacyl-type photoinitiator based on ethyl carbazole as a long wavelength photo-initiator is developed for free radical polymerization. Phenacyl ethyl carbazolium hexafluoroantimonate (PECH) photoinitiator is synthesized in a two-step, one-pot manner by quaternizing ethyl carbazole with phenacyl bromide and
Ceren Kütahya et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(46), 10444-10451 (2020-04-29)
NIR exposure at 790 nm activated photopolymerization of monomers comprising UV-absorbing moieties by using [CuII /(TPMA)]Br2 (TPMA=tris(2-pyridylmethyl)amine) in the ppm range and an alkyl bromide as initiator. Some of them comprised structural elements selected either from those showing proton transfer or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service