Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

530506

Sigma-Aldrich

Dichloromethane-d2

99.9 atom % D, contains 0.1 % (v/v) TMS

Synonym(s):

Dideuteromethylenechloride, Methylene chloride-d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD2Cl2
CAS Number:
Molecular Weight:
86.94
Beilstein:
1733318
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.9 atom % D

Assay

≥99.00% (GC)

form

liquid

contains

0.1 % (v/v) TMS

packaging

ampule of 1g
glass bottle of 25g

technique(s)

NMR: suitable

impurities

≤0.0100% water
water

refractive index

n20/D 1.422 (lit.)

bp

40 °C (lit.)

mp

-97 °C (lit.)

density

1.362 g/mL at 25 °C (lit.)

mass shift

M+2

SMILES string

[2H]C([2H])(Cl)Cl

InChI

1S/CH2Cl2/c2-1-3/h1H2/i1D2

InChI key

YMWUJEATGCHHMB-DICFDUPASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichloromethane-d2 (CD2Cl2) is a deuterated NMR solvent containing 0.1% (v/v) TMS (tetramethylsilane). It is useful in NMR-based research and analyses. Raman spectral analysis of CD2Cl2 in the gas phase has been reported.

Application

Dichloromethane-d2 may be used to analyze the three-dimensional structure of (9S,11S)-11-amino-9-deoxo-11,12-deoxy-9,12-epoxyerythromycin by proton NMR derived constrained molecular mechanics.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of the three-dimensional, solution-phase structure of the macrolide antibiotic oxolide in CD2Cl2 and D2O from NMR constraints.
Steinmetz WE, et al.
Magnetic Resonance in Chemistry, 43(1), 16-20 (2005)
Absolute Raman intensities, force constants, and electro-optical parameters of CH2Cl2, CD2Cl2 and CHDCl2.
Escribano R, et al.
Molecular Physics, 37(2), 361-377 (1979)
Ting Li et al.
Environmental science and pollution research international, 22(15), 11792-11800 (2015-04-11)
We surveyed the occurrence of nine N-nitrosamine species in ten bottled drinking waters from supermarket and other water samples including raw waters, finished waters, and distribution system waters from nine municipal drinking water treatment plants in eight cities of Jiangsu
Alejandro G Lichtscheidl et al.
Dalton transactions (Cambridge, England : 2003), 44(36), 16156-16163 (2015-08-22)
The synthesis and full characterization, including Nuclear Magnetic Resonance (NMR) data ((1)H, (13)C{(1)H} and (119)Sn{(1)H}), for a series of Me3SnX (X = O-2,6-(t)Bu2C6H3 (), (Me3Sn)N(2,6-(i)Pr2C6H3) (), NH-2,4,6-(t)Bu3C6H2 (), N(SiMe3)2 (), NEt2, C5Me5 (), Cl, Br, I, and SnMe3) compounds in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service