Skip to Content
MilliporeSigma
All Photos(2)

Documents

441406

Sigma-Aldrich

Tetrahydrofuran-d8

≥99.5 atom % D

Synonym(s):

THF-d8, Deuterated tetrahydrofuran, Octadeuterotetrahydrofuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4D8O
CAS Number:
Molecular Weight:
80.16
Beilstein:
111854
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

≥99.5 atom % D

Quality Level

Assay

≥99% (CP)

form

liquid

technique(s)

NMR: suitable

impurities

≤0.03% water
water

refractive index

n20/D 1.403 (lit.)

bp

65-66 °C (lit.)

mp

−106 °C (lit.)

density

0.985 g/mL at 25 °C (lit.)

mass shift

M+8

storage temp.

2-8°C

SMILES string

[2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2/i1D2,2D2,3D2,4D2

InChI key

WYURNTSHIVDZCO-SVYQBANQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrofuran-d8 (THF-d8, TDF) is a deuterated NMR solvent useful in NMR-based research and analyses. 1H NMR line shape analysis of the exchange rates of proton between methanol and acetic acid dissolved in THF-d8 was reported. It participates as a solvent in the synthesis of deuteriotris[bis(trimethylsilyl)amido]thorium and deuteriotris[bis(trimethylsiiyl)amido]uranium. Dynamic NMR spectroscopic study of 15N,15N′-dipentadeuterophenylformamidine (DPFA) has been studied in THF-d8.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and hydrogen-deuterium exchange of alkyl and hydride bis(trimethylsilyl) amido derivatives of the actinide elements.
Simpson SJ, et al.
Inorganic Chemistry, 20(9), 2991-2995 (1981)
NMR lineshape theory of superimposed intermolecular spin exchange reactions and its action to the system acetic acid/methanol/tetrahydrofuran-d8.
Limbach HH.
Journal of Magnetic Resonance, 36(3), 287-300 (1979)
Dynamic NMR study of the interference between cyclic proton exchange, selfassociation and hindered rotation of diphenylformamidine in tetrahydrofuran.
Meschede L, et al.
Berichte der Bunsengesellschaft fur physikalische Chemie, 92(4), 469-485 (1988)
Alejandro G Lichtscheidl et al.
Dalton transactions (Cambridge, England : 2003), 44(36), 16156-16163 (2015-08-22)
The synthesis and full characterization, including Nuclear Magnetic Resonance (NMR) data ((1)H, (13)C{(1)H} and (119)Sn{(1)H}), for a series of Me3SnX (X = O-2,6-(t)Bu2C6H3 (), (Me3Sn)N(2,6-(i)Pr2C6H3) (), NH-2,4,6-(t)Bu3C6H2 (), N(SiMe3)2 (), NEt2, C5Me5 (), Cl, Br, I, and SnMe3) compounds in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service