Skip to Content
MilliporeSigma
All Photos(1)

Documents

522929

Sigma-Aldrich

3,5-Dimethylbenzenethiol

90%

Synonym(s):

3,5-Dimethylthiophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3SH
CAS Number:
Molecular Weight:
138.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

refractive index

n20/D 1.568 (lit.)

bp

127.5 °C/50 mmHg (lit.)

density

1.015 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)cc(S)c1

InChI

1S/C8H10S/c1-6-3-7(2)5-8(9)4-6/h3-5,9H,1-2H3

InChI key

CESBAYSBPMVAEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,5-Dimethylbenzenethiol is an organic building block. It is also referred to as 5-mercapto-m-xylene.

Application

3,5-Dimethylbenzenethiol may be employed as a starting reagent in the preparation of Lithium 2-lithio-3,5-dimethylbenzenethiolate. It may also be used in the preparation of 1,1,2-trichloro-4-(3,5-dimethylphenylthio)-1-buten-3-yne and 1,1,2,4-tetrachloro-4-(3,5-dimethylphenylthio)-1,3-butadiene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Directed lithiation of arenethiols.
Smith K, et al.
Journal of the American Chemical Society, 111(2), 665-669 (1989)
Ibis C and Sahin A.
Bull. Korean Chem. Soc., 31(8), 2255-2260 (2010)
Homan Kang et al.
Scientific reports, 5, 10144-10144 (2015-05-29)
Recently, preparation and screening of compound libraries remain one of the most challenging tasks in drug discovery, biomarker detection, and biomolecular profiling processes. So far, several distinct encoding/decoding methods such as chemical encoding, graphical encoding, and optical encoding have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service