Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

518654

Sigma-Aldrich

4-Amino-2-chloro-6,7-dimethoxyquinazoline

95%

Synonym(s):

2-Chloro-6,7-dimethoxy-4-quinazolinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10ClN3O2
CAS Number:
Molecular Weight:
239.66
Beilstein:
747463
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

262-268 °C (dec.) (lit.)

SMILES string

COc1cc2nc(Cl)nc(N)c2cc1OC

InChI

1S/C10H10ClN3O2/c1-15-7-3-5-6(4-8(7)16-2)13-10(11)14-9(5)12/h3-4H,1-2H3,(H2,12,13,14)

InChI key

HWIIAAVGRHKSOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V A Alabaster et al.
Journal of medicinal chemistry, 30(6), 999-1003 (1987-06-01)
A series of 4-amino-2-(4-carbamoylpiperidino)-6,7-dimethoxyquinazolines (2) were synthesized and evaluated for alpha-adrenoceptor affinity and antihypertensive activity. These compounds displayed high binding affinity (Ki, 10(-9)-10(-10) M) and selectivity for alpha 1-adrenoceptors in vitro, and 12, 14, 21-26, and 29 showed similar potency
S F Campbell et al.
Journal of medicinal chemistry, 30(10), 1794-1798 (1987-10-01)
A series of 4-amino-6,7-dimethoxy-2(4-heterocyclylpiperazin-1-yl)quinazolines (3) was prepared and screened for alpha-adrenoceptor affinity and antihypertensive activity. These quinazoline derivatives showed high binding affinity (ca. 10(-10) M) and selectivity (greater than 10,000) for alpha 1-adrenoceptors in vitro, with no relevant activity at
D Giardinà et al.
Journal of medicinal chemistry, 39(23), 4602-4607 (1996-11-08)
The enantiomers of [4-(4-amino-6, 7-dimethoxyquinazolin-2-yl)-cis-octahydroquinoxalin-1-yl]-fu ran- 2-ylmethanone (cyclazosin, 1) were synthesized from the chiral furan-2-yl(cis-octahydroquinoxalin-1-yl)methanone [(+)-2 and (-)-2], which were obtained by resolution of the racemic amine with (S)-(+)- and (R)-(-)-mandelic acid. The binding profile of the enantiomers of 1
A Leonardi et al.
Journal of medicinal chemistry, 42(3), 427-437 (1999-02-13)
A new series of novel piperazine and non-piperazine derivatives of 2, 4-diamino-6,7-dimethoxyquinazoline was synthesized and evaluated for binding affinity toward alpha1-adrenergic and other G-protein-coupled aminergic receptors. The alpha1-adrenoceptor (AR) subtype selectivity was also investigated for the most interesting compounds. Only

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service