Skip to Content
MilliporeSigma
4-Amino-2-chloro-6,7-dimethoxyquinazoline 95%

4-Amino-2-chloro-6,7-dimethoxyquinazoline

4-Amino-2-chloro-6,7-dimethoxyquinazoline 95%
Synonym(s):
2-Chloro-6,7-dimethoxy-4-quinazolinamine
Empirical Formula (Hill Notation):
C10H10ClN3O2
CAS Number:
Molecular Weight:
239.66
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing