Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

515051

Sigma-Aldrich

Potassium ethoxide solution

24 wt. % in ethanol

Synonym(s):

Potassium ethanolate, Potassium ethylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOC2H5
CAS Number:
Molecular Weight:
84.16
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

concentration

24 wt. % in ethanol

refractive index

n20/D 1.39

density

0.894 g/mL at 25 °C

SMILES string

[K+].CC[O-]

InChI

1S/C2H5O.K/c1-2-3;/h2H2,1H3;/q-1;+1

InChI key

RPDAUEIUDPHABB-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Self-heat. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

70.0 °F - closed cup

Flash Point(C)

21.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service