Skip to Content
MilliporeSigma
All Photos(1)

Documents

514691

Sigma-Aldrich

Ethyl 4-cyanobenzoylformate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4COCO2C2H5
CAS Number:
Molecular Weight:
203.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

refractive index

n20/D 1.537 (lit.)

bp

313 °C (lit.)

density

1.18 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(=O)c1ccc(cc1)C#N

InChI

1S/C11H9NO3/c1-2-15-11(14)10(13)9-5-3-8(7-12)4-6-9/h3-6H,2H2,1H3

InChI key

MGBZXMQPYAYPIQ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regina Kratzer et al.
Microbial cell factories, 7, 37-37 (2008-12-17)
Whole cell-catalyzed biotransformation is a clear process option for the production of chiral alcohols via enantioselective reduction of precursor ketones. A wide variety of synthetically useful reductases are expressed heterologously in Escherichia coli to a high level of activity. Therefore
Christoph Gruber et al.
Biotechnology journal, 8(6), 699-708 (2013-04-17)
Generally, recombinant and native microorganisms can be employed as whole-cell catalysts. The application of native hosts, however, shortens the process development time by avoiding multiple steps of strain construction. Herein, we studied the NAD(P)H-dependent reduction of o-chloroacetophenone by isolated xylose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service