Skip to Content
MilliporeSigma
All Photos(1)

Documents

CRM40570

Supelco

DBA-d9 Monomers Internal Standard Mix

certified reference material, in acetone (varied concentration), ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

in acetone (varied concentration)

technique(s)

gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    ICA-DBA-D9 DERIVATIVE
    MIC-DBA-D9-DERIVATIVE
    EIC-DBA-D9 DERIVATIVE
    PIC-DBA-D9 DERIVATIVE
    PHI-DBA-D9 DERIVATIVE
    1,6-HDI-DBA-D18 DERIVATIVE
    2,4-TDI-DBA-D18 DERIVATIVE
    2,6-TDI-DBA-D18 DERIVATIVE
    4,4-MDI-DBA-D18 DERIVATIVE
    IPDI-DBA-D18 DERIVATIVE ISOMER 1
    IPDI-DBA-D18 DERIVATIVE ISOMER 2
    See All (11)

sample collection

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Per Leanderson et al.
Archives of environmental & occupational health, 75(3), 159-164 (2019-05-10)
Pyrolysis of nitrogen containing biofuels generates isocyanic acid (ICA) and we here studied if ICA also is present in cauterization smoke. Air sampling was performed when animal technicians that had developed airway symptoms worked with dehorning. Tissue heated in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service