512559
Bis(isopropylcyclopentadienyl)tungsten(IV) dihydride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C5H4CH(CH3)2)2WH2
CAS Number:
Molecular Weight:
400.20
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
liquid
composition
W, 43.8-48.0% gravimetric (with Cinchonine)
reaction suitability
core: tungsten
reagent type: catalyst
refractive index
n20/D 1.6 (lit.)
bp
230 °C (lit.)
SMILES string
[WH2].CC(C)[C]1[CH][CH][CH][CH]1.CC(C)[C]2[CH][CH][CH][CH]2
InChI
1S/2C8H11.W.2H/c2*1-7(2)8-5-3-4-6-8;;;/h2*3-7H,1-2H3;;;
InChI key
BRZLVTGJFZKZNZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
accessory
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids not in Storage Class 3
WGK
WGK 3
Flash Point(F)
199.9 °F - closed cup
Flash Point(C)
93.3 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service