Skip to Content
MilliporeSigma
All Photos(1)

Documents

510807

Sigma-Aldrich

Bis(cyclopentadienyl)tungsten(IV) dihydride

97%

Synonym(s):

Bis(η5 -cyclopentadienyl)tungsten dihydride, Bis(cyclopentadienyl)dihydrotungsten, Dicyclopentadienyltungsten dihydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12W
CAS Number:
Molecular Weight:
316.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder or crystals

reaction suitability

core: tungsten
reagent type: catalyst

mp

115 °C (lit.)

SMILES string

[H][W][H].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2

InChI

1S/2C5H5.W.2H/c2*1-2-4-5-3-1;;;/h2*1-5H;;;

InChI key

FCGFDFIKNFHEHZ-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service