43761
2,2′-Dithiodibenzoic acid
≥95.0% (NT)
Synonym(s):
2-Carboxyphenyl disulfide, Bis(2-carboxyphenyl) disulfide, Dithiosalicylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
S2(C6H4CO2H)2
CAS Number:
Molecular Weight:
306.36
Beilstein:
2221810
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥95.0% (NT)
ign. residue
≤3%
mp
287-290 °C (lit.)
288-293 °C
functional group
carboxylic acid
disulfide
SMILES string
OC(=O)c1ccccc1SSc2ccccc2C(O)=O
InChI
1S/C14H10O4S2/c15-13(16)9-5-1-3-7-11(9)19-20-12-8-4-2-6-10(12)14(17)18/h1-8H,(H,15,16)(H,17,18)
InChI key
LBEMXJWGHIEXRA-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
2,2′-Dithiodibenzoic acid is an sulfhydryl modifying reagent. 2,2′-Dithiodibenzoic acid on cocrystallization with imidazole or 4-methylimidazole affords bis(imidazolium) 2,2′-dithiodibenzoate and 4-methylimidazolium 2-[(2-carboxyphenyl)disulfanyl]benzoate organic salt, respectively. It also cocrystallizes with isonicotinohydrazide from methanol solution to afford the 1:2 cocrystal, 2,2′-dithiodibenzoic acid-isonicotinohydrazide.
Application
2,2′-Dithiodibenzoic acid may be used for the preparation of novel anionic heptadecanuclear silver(I) cluster, by the reaction with equivalent molar silver oxide under ultrasonic conditions at 50°C. It may be used for the preparation of polyamides containing a disulfide bond in their main chain.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
New type polyamides containing disulfide bonds for positive active material of lithium secondary batteries.J. power
Tsutsumi H, et al.
Journal of Power Sources, 68(2), 735-738 (1997)
Xiang-Gao Meng et al.
Acta crystallographica. Section C, Crystal structure communications, 64(Pt 5), o261-o263 (2008-05-03)
Cocrystallization of 2,2'-dithiodibenzoic acid with isonicotinohydrazide from methanol solution yields the 1:2 cocrystal 2,2'-dithiodibenzoic acid-isonicotinohydrazide (1/2), C(14)H(10)O(4)S(2) x 2 C(6)H(7)N(3)O. The component molecules are linked by intermolecular O-H...N, N-H...O, N-H...N and C-H...O hydrogen bonds into layers running parallel to the
Di Sun et al.
Inorganic chemistry, 50(24), 12393-12395 (2011-11-11)
A novel anionic heptadecanuclear silver(I) cluster, (NH(4))(17)[(μ(6)-S)@Ag(17)(mba)(16)]·22H(2)O (1; H(2)mba = 2-mercaptobenzoic acid), was obtained by the reaction of equivalent molar silver oxide and 2,2'-dithiodibenzoic acid (H(2)dtba) under ultrasonic conditions at 50 °C. Complex 1 is a discrete cluster comprised of
J E Bodwell et al.
Biochemistry, 23(7), 1392-1398 (1984-03-27)
Glucocorticoid -receptor complexes from intact rat thymus cells incubated with [3H]dexamethasone at 0 degree C are in the nonactivated form and do not bind to DNA-cellulose. Upon being warmed, they are transformed to activated complexes that bind to DNA-cellulose at
Linheng Wei et al.
Acta crystallographica. Section C, Crystal structure communications, 65(Pt 9), o453-o456 (2009-09-04)
Cocrystallization of imidazole or 4-methylimidazole with 2,2'-dithiodibenzoic acid from methanol solution yields the title 2:1 and 1:1 organic salts, 2C(3)H(5)N(2)(+).C(14)H(10)O(4)S(2)(2-), (I), and C(4)H(7)N(2)(+).C(14)H(10)O(4)S(2)(-), (II), respectively. Compound (I) crystallizes in the monoclinic C2/c space group with the mid-point of the S-S
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service