Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50920

Sigma-Aldrich

Guanidine acetate salt

≥99%

Synonym(s):

Guanidinium monoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NH2 · CH3COOH
CAS Number:
Molecular Weight:
119.12
Beilstein:
3565247
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

powder

solubility

H2O: 0.1 g/mL, clear

functional group

amine
carboxylic acid

SMILES string

CC(O)=O.NC(N)=N

InChI

1S/C2H4O2.CH5N3/c1-2(3)4;2-1(3)4/h1H3,(H,3,4);(H5,2,3,4)

InChI key

DXTIKTAIYCJTII-UHFFFAOYSA-N

General description

Guanidine acetate salt is also known as guanidinium acetate. Its TG (thermogravimetry) and DSC (differential scanning calorimetry) curves have been reported.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A TG-DSC investigation of the thermal dissociation of selected guanidinium salts.
Wendlandt WW, et al.
Thermochimica Acta, 75(1), 239-244 (1984)
Shan Xu et al.
Cellular and molecular biology (Noisy-le-Grand, France), 66(5), 125-130 (2020-10-13)
Diarrhea is a disease, and patients must have bowel movements at least three times per day. This condition may last for several days and may cause dehydration due to fluid loss. Spleen-deficiency makes a person more vulnerable to some infectious

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service