Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

499307

Sigma-Aldrich

Phenethylzinc bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2ZnBr
CAS Number:
Molecular Weight:
250.45
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

concentration

0.5 M in THF

bp

65 °C

density

0.965 g/mL at 25 °C

functional group

phenyl

storage temp.

2-8°C

SMILES string

Br[Zn]CCc1ccccc1

InChI

1S/C8H9.BrH.Zn/c1-2-8-6-4-3-5-7-8;;/h3-7H,1-2H2;1H;/q;;+1/p-1

InChI key

SEXSOTWHLBYVNF-UHFFFAOYSA-M

Application

Phenethylzinc bromide can be used as a reagent:
  • In the transition metal-catalyzed Negishi cross-coupling reaction to prepare aryl or heteroaryl derivatives by reacting with organic halides or triflates via carbon-carbon bond formation.
  • To prepare (4-phenyl-1-butyn-1-yl)benzene by reacting with phenylacetylene via palladium-catalyzed cross-coupling reaction.
  • To synthesize benzyl substituted thienopyrimidine derivatives.

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed aerobic oxidative cross-coupling reactions of terminal alkynes with alkylzinc reagents
Chen M, et al.
Journal of the American Chemical Society, 132(12), 4101-4103 (2010)
Non-peptidic small molecule inhibitors of XIAP
Park C-M, et al.
Bioorganic & Medicinal Chemistry Letters, 15(3), 771-775 (2005)
Substituted thieno [2, 3-d] pyrimidines as adenosine A2A receptor antagonists
Shook BC, et al.
Bioorganic & Medicinal Chemistry Letters, 23(9), 2688-2691 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service