Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

499110

Sigma-Aldrich

4-Chlorobenzaldehyde dimethyl acetal

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH(OCH3)2
CAS Number:
Molecular Weight:
186.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.508 (lit.)

bp

114-115 °C/19 mmHg (lit.)

density

1.128 g/mL at 25 °C (lit.)

functional group

acetal
chloro
ether

SMILES string

COC(OC)c1ccc(Cl)cc1

InChI

1S/C9H11ClO2/c1-11-9(12-2)7-3-5-8(10)6-4-7/h3-6,9H,1-2H3

InChI key

YRNBYTFFWMWTGG-UHFFFAOYSA-N

General description

4-Chlorobenzaldehyde dimethyl acetal has been prepared from 4-chlorobenzaldehyde in the presence of cerium (lV) ammonium nitrate and characterized by 1H and 13C-NMR.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A mild and efficient procedure for the preparation of dimethyl and diallyl acetals of aromatic aldehydes mediated by cerium (lV) ammonium nitrate.
Nair V, et al.
Indian J. Chem. B, 44(1), 141-143 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service