Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

494208

Sigma-Aldrich

N-Methyl-p-toluidine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NHCH3
CAS Number:
Molecular Weight:
121.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.557 (lit.)

bp

212 °C (lit.)

density

0.958 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CNc1ccc(C)cc1

InChI

1S/C8H11N/c1-7-3-5-8(9-2)6-4-7/h3-6,9H,1-2H3

InChI key

QCIFLGSATTWUQJ-UHFFFAOYSA-N

General description

N-Methyl-p-toluidine is an aromatic secondary amine. It forms social isomers when encapsulated with a molecule of chloroform or benzene in a cylindrical capsule. 2-amino-4,6-dichloropyrimidine-5-carbaldehyde undergoes mono-amination with N-methyl-p-toluidine to form a precursor for the preparation of pyrazolo[3,4-d]pyrimidines. It annelates with dimethyl acetylenedicarboxylate via formation of toluidine radical cation intermediate.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

183.0 °F - closed cup

Flash Point(C)

83.89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous encapsulation: molecules held at close range.
Rebek J.
Angewandte Chemie (International Edition in English), 44(14), 2068-2078 (2005)
Microwave-assisted synthesis of pyrazolo [3,4-d] pyrimidines from 2-amino-4, 6-dichloropyrimidine-5-carbaldehyde under solvent-free conditions.
Quiroga J, et al.
Tetrahedron Letters, 49(20), 3257-3259 (2008)
Palladium-promoted aromatic annelation with acetylene dicarboxylates.
Sakakibara T, et al.
Chemistry Letters (Jpn), 15(5), 797-800 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service