Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

492841

Sigma-Aldrich

2-Propanol-2-d1

98 atom % D

Synonym(s):

Isopropanol-2-d1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CDOH
CAS Number:
Molecular Weight:
61.10
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Assay

99% (CP)

refractive index

n20/D 1.377 (lit.)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

bulk density

0.785 g/mL

mass shift

M+1

SMILES string

[2H]C(C)(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3/i3D

InChI key

KFZMGEQAYNKOFK-WFVSFCRTSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service