Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

488313

Sigma-Aldrich

Butane-1,4-13C2

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3CH2CH213CH3
CAS Number:
Molecular Weight:
60.11
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

vapor density

2.11 (vs air)

vapor pressure

51.6 psi ( 37.7 °C)

isotopic purity

99 atom % 13C

bp

−0.5 °C (lit.)

mp

−138 °C (lit.)

mass shift

M+2

SMILES string

[13CH3]CC[13CH3]

InChI

1S/C4H10/c1-3-4-2/h3-4H2,1-2H3/i1+1,2+1

InChI key

IJDNQMDRQITEOD-ZDOIIHCHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Brass hose adapter Z146811 or brass body mini gas regulator Z513539 is recommended.

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-76.0 °F - closed cup

Flash Point(C)

-60 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service