Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

724998

Sigma-Aldrich

1-Butanol-13C4

99 atom % 13C

Synonym(s):

Butyl-13C4 alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3(13CH2)3OH
CAS Number:
Molecular Weight:
78.09
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

liquid

bp

116-118 °C (lit.)

density

0.853 g/mL at 25 °C

mass shift

M+4

SMILES string

[13CH3][13CH2][13CH2][13CH2]O

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3/i1+1,2+1,3+1,4+1

InChI key

LRHPLDYGYMQRHN-JCDJMFQYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service