Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

488291

Sigma-Aldrich

Bromomethane-d3

99.5 atom % D

Synonym(s):

Methyl-d3 bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3Br
CAS Number:
Molecular Weight:
97.96
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

vapor density

3.3 (20 °C, vs air)

Quality Level

vapor pressure

1420 mmHg ( 20 °C)

isotopic purity

99.5 atom % D

autoignition temp.

996 °F

bp

4 °C (lit.)

mp

−94 °C (lit.)

density

3.41 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

mass shift

M+3

SMILES string

[2H]C([2H])([2H])Br

InChI

1S/CH3Br/c1-2/h1H3/i1D3

InChI key

GZUXJHMPEANEGY-FIBGUPNXSA-N

General description

Bromomethane-d3 (CD3Br) is a perdeuterated isotopomer of methyl bromide. Its vibrationless Rydberg transitions have been investigated based on photoabsorption spectral data.[1] The infrared spectrum of CD3Br has been analyzed in the range 2000-7000cm-1.[2]

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Shipping of this product by air freight is forbidden.
Outside of the U.S., ocean freight shipment is required and will result in substantial additional expense.
Standard 10 g quantity is packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressure at 21ºC is 13 psig for 10 g. This pressure is slightly above atmospheric pressure. Care must be taken when extracting product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951 or Z187224 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Gas 2 - Muta. 2 - Ozone 1 - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joint local-and normal-mode studies of the overtone spectra of the methyl halides: CH3F, CH3Cl, CH3Br, CD3Br, and CH3I.
Law MM.
J. Chem. Phys. , 111(22), 10021-10033 (1999)
The vacuum UV photoabsorption spectrum of methyl bromide (CH3Br) and its perdeuterated isotopomer CD3Br: a Rydberg series analysis.
Locht R, et al.
Chemical Physics, 317(1), 73-86 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service