Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

486221

Sigma-Aldrich

Trimethyl-d9-amine

99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)3N
CAS Number:
Molecular Weight:
68.17
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

vapor density

2.09 (vs air)

Quality Level

vapor pressure

13.3 psi ( 21 °C)

isotopic purity

99 atom % D

autoignition temp.

374 °F

bp

3-4 °C (lit.)

mp

−117 °C (lit.)

density

0.734 g/mL at 25 °C (lit.)

mass shift

M+9

SMILES string

[2H]C([2H])([2H])N(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C3H9N/c1-4(2)3/h1-3H3/i1D3,2D3,3D3

InChI key

GETQZCLCWQTVFV-GQALSZNTSA-N

Packaging

5 g quantity is packaged in a 450 mL carbon steel lecture bottle with stainless steel CGA 110/180 valve. Nominal gas pressure at 21ºC is 13 psig for 5 g. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951, Z187224 or either of stainless steel regulators Z148504 and Z148512 and a "T"-purge valve Z151661 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Gas 1A - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service