Skip to Content
MilliporeSigma
All Photos(1)

Documents

485489

Sigma-Aldrich

Dichloroacetic acid-2-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl213CHCO2H
CAS Number:
Molecular Weight:
129.93
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

bp

194 °C (lit.)

mp

9-11 °C (lit.)

density

1.575 g/mL at 25 °C

mass shift

M+1

SMILES string

OC(=O)[13CH](Cl)Cl

InChI

1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)/i1+1

InChI key

JXTHNDFMNIQAHM-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Acute 1 - Carc. 2 - Eye Dam. 1 - Lact. - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A - STOT RE 2 Oral

Target Organs

Brain,Liver,Testes

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shimin Wu et al.
Journal of chromatography. A, 1487, 100-107 (2017-01-26)
The addition of oxidants for disinfecting water can lead to the formation of potentially carcinogenic compounds referred to as disinfection byproducts (DBPs). Haloacetic acids (HAAs) are one of the most widely detected DBPs in US water utilities and some of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service