Skip to Content
MilliporeSigma

604232

Dichloroacetic acid-1-13C

99 atom % 13C, 97% (CP)

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
Cl2CH13CO2H
CAS Number:
Molecular Weight:
129.93
UNSPSC Code:
12352101
PubChem Substance ID:
MDL number:
Isotopic purity:
99 atom % 13C
Assay:
97% (CP)
Mass shift:
M+1


isotopic purity

99 atom % 13C

assay

97% (CP)

bp

194 °C (lit.)

mp

9-11 °C (lit.)

density

1.575 g/mL at 25 °C

mass shift

M+1

SMILES string

O[13C](=O)C(Cl)Cl

InChI

1S/C2H2Cl2O2/c3-1(4)2(5)6/h1H,(H,5,6)/i2+1

InChI key

JXTHNDFMNIQAHM-VQEHIDDOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Acute 1 - Carc. 2 - Eye Dam. 1 - Lact. - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A - STOT RE 2 Oral

target_organs

Brain,Liver,Testes

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

235.4 °F

flash_point_c

113 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library