Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

485187

Sigma-Aldrich

1-Bromohexane-d13

98 atom % D

Synonym(s):

Hexyl-d13 bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3C(CD2)4CD2Br
CAS Number:
Molecular Weight:
178.15
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

refractive index

n20/D 1.4475 (lit.)

bp

154-158 °C (lit.)

mp

−85 °C (lit.)

density

1.269 g/mL at 25 °C

mass shift

M+13

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])Br

InChI

1S/C6H13Br/c1-2-3-4-5-6-7/h2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2

InChI key

MNDIARAMWBIKFW-UTBWLCBWSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caitlyn M Wolf et al.
Soft matter, 15(25), 5067-5083 (2019-06-12)
The molecular morphology and dynamics of conjugated polymers in the bulk solid state play a significant role in determining macroscopic charge transport properties. To understand this relationship, molecular dynamics (MD) simulations and quantum mechanical calculations are used to evaluate local

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service